Zentrifugenröhrchen, Glas, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
216011004 216011107 216011407 216011707 216012403 216012609 216013605 216111705 242630901 216111405
525-1102EA 7.9 EUR InStock
525-1102 525-1104 525-1106 525-1108 525-1110 525-1112 525-1114 525-1142 525-1122 525-1132
Zentrifugenröhrchen, Glas, DURAN®
Röhrchen Zentrifugenröhrchen
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical and thermal resistance
  • Max. RCF: 4000×g
Robust universal centrifuge tubes for a wide variety of applications.
525-1110 and 525-1114 not in accordance with DIN.
Zertifizierungen: DIN 58970 (Part 2) only for round bottom tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge