Organic reference standard, Eupatorin, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
52559-10MG
52559-10MGEA 230 EUR InStock
52559-10MG
Organic reference standard, Eupatorin, analytical standard, Supelco®
Standards Organische Standards
Eupatorin acts as an anti-proliferative in the cells expressing the CYP1A- family. It induces G2/M block followed by apoptosis in cells expressing the CYP1A- family. It also functions as an anti-inflammatory.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3632d8b442365d0a815c8fc170ae6eb5c46be9eb&vid=3632d8b442365d0a815c8fc170ae6eb5c46be9eb&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge