Sauerstoffflaschen nach Winkler
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
05605049 05605037
528-1203EA 33.5 EUR InStock
528-1203 528-1205
Sauerstoffflaschen nach Winkler
Flaschen Sauerstoffflaschen
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge