Rotary evaporator, Multivapor™ P-12
Lieferant: BUCHI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MP11001R00 MP11001S10
531-0185EA 9810 EUR InStock
531-0185 531-0186
Rotary evaporator, Multivapor™ P-12
Verdampfer Rotationsverdampfer
Compact device for parallel vacuum evaporation of up to 12 samples. Used in conjunction with a Rotavapor®, this enhances performance by transferring vapours to the condenser of the rotary evaporator; used alone, it offers an alternative independent solution.
  • Time saving: 12 samples simultaneously
  • Reduced footprint: 260×400 mm
  • No cross-contamination thanks to the individual sealing of each channel
Multivapor P-12 Rotavapor: for use in conjunction with an existing rotary evaporator. Includes device with connections for condenser, no condenser
Multivapor P-12 Basic: complete standalone system with plastic-coated glassware including a 1000-ml KS 24/20 receiving flask, no pump, no controller
Lieferumfang: Supplied with 15 tubes and adaptors.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge