Vacuum concentrators, refrigerated, CentriVap®
Lieferant: LABCONCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7310030 7310031 7310035 7310036 7310032 7310034
531-0189EA 13610 EUR InStock
531-0189 531-0190 LBCP7310035 531-0998 531-1030 LBCP7310034
Vacuum concentrators, refrigerated, CentriVap®
Verdampfer Zentrifugalverdampfer
The refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature. The concentrator combines a refrigeration system that cools to −4 °C with a heater that warms to 100 °C to achieve the user's desired set point temperatures. With an epoxy-coated aluminum chamber.
Vacuum pumps provide a reliable vacuum source for the concentrators.
CentriVap Rotors hold tubes at an angle to maximize the surface area and are easily interchangeable. Glassware is not included.
Traps are connected between the evaporator and vacuum pump to provide additional protection to the pump. The inserts require a clear polypropylene canister with mounting bracket or clear polypropylene canister with stand for proper use.
Bestellinformation: Other rotors and traps are available. Please contact your Avantor customer service team for further information.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge