Recirculating chillers, F-100 / F-108 / F-114 / F-125
Lieferant: BUCHI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11056460 11056464
531-0858EA 3170 EUR InStock
531-0858 531-0923
Recirculating chillers, F-100 / F-108 / F-114 / F-125
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
The F-series chillers provide the final step to a fully integrated evaporation or distillation process, leading to several key benefits:
  • Up to 75% less energy consumption
  • Reduced heat emission
  • Easy handling and mobility
  • Optimised distillation capacity
  • Increased process safety due to communication of all connected instruments
F-100: Cost effective, small recirculating chiller that fulfils the basic cooling requirements. The perfect replacement of tap water chilling with small distillation instruments. It cools to a fixed temperature of 10 °C and is ideal for use with one rotary evaporator.
F-108: Powerful alternative to the F-100. The chiller is capable of cooling to −10 °C. At 15 °C the F-108 provides 800 W cooling capacity. If connected to one rotary evaporator it is possible to keep the temperature at 0 °C when performing a normal distillation.
F-114: Ideal when very low temperatures are required for single or multiple units. At 15 °C the F-114 provides a powerful 1400 W cooling capacity and ultimately cools to a final temperature of −10 °C.
F-125: A powerful industrial chiller with a cooling capacity of 2500 W at 15 °C and an integrated trolley for use with the Rotavapor® R-220 SE industrial rotary evaporator.
F-105: is environmentally friendly and compact recirculation chiller designed for one laboratory evaporator. This features an adjustable temperature with a higher cooling capacity. In addition, the F-105 offers ON/OFF regulation when used in conjunction with the Interface. Cooling capacity of 530 W at 15 °C, 50/60 Hz frequency.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge