Recombinant protein G, peroxidase conjugated, Calbiochem®
Lieferant: Novabiochem (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
539322-500
539322-500EA 234 EUR InStock
539322-500
Recombinant protein G, peroxidase conjugated, Calbiochem®
Protein-Aufbereitungs Proteinreinigungs-Reagenzien
Purified recombinant Protein G conjugated to peroxidase can be used in direct immunoassay.
  • Used in enzyme-linked immunosorbent assay and immunoblotting (Western blotting)
  • Binds ≥99% to human IgG immobilised on a column
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge