VWR®, Vial Racks
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
548-0131AEA 119 EUR InStock
548-0131A 548-1520A 548-0192A
VWR®, Vial Racks
Gestelle Gestelle für Chromatographie-Ampullen
Vial Racks are ideal for storage and transportation of vials.
  • Safe storage of vials
  • Stable construction
  • Easy handling and transportation
These vial racks allow the vials to stand securely on the laboratory bench. The empty racks are stackable and therefore space-saving.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge