VWR® ND11, Rollrandflaschen
Lieferant: LA PHA PACK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11090476 11090356 11090477 11090619
548-3004EA 30.1 EUR InStock
548-3004 548-3003 548-3005 548-3092 548-0906 548-1362 548-2126 548-2159
VWR® ND11, Rollrandflaschen
Vials
Diese Flaschen sind für Instrumente der folgenden Hersteller geeignet: Agilent, Carlo Erba, CTC, DANI, Fisons, Gerstel, Jasco, PerkinElmer, Shimadzu, Spark, Thermo Scientific, Varian etc.
  • Standard vials for GC and HPLC
  • Vials with a barcode label, pre-crimped vials are also available
  • Microlitre vials for sample preparation (reactions, concentrations) or as an alternative for conical micro-vials or crimp neck vials with inserts
TopSert vials are an alternative to glass vials with fused-in micro-inserts or micro-insert with plastic spring. The TopSert glass micro-insert is centred in the plastic mould and pressed firmly against the septa due to its slightly overlapping edge.
Lieferumfang: Sets enthalten: 1,5 ml Rollrandflaschen (11,6×32 mm), Bördelkappe aus Aluminium, klar lackiert, mit zentralem Loch (5,5 mm) und Septum.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge