Needle cleaning concentrate and wires
Lieferant: HAMILTON BONADUZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
18311 76620 18310 18300 18301 18302 18303 18304 18306
549-1250EA 104 EUR InStock
549-1250 549-1251 549-1249 549-1244 549-1245 549-1246 549-1247 549-1248 HAMI18306
Needle cleaning concentrate and wires
Reinigungsmittel Waschmittel und Oberflächenreiniger
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally the solvent would be non-alkaline, non-phosphate, and non-detergent based.
  • Biodegradable, non-phosphate, cleaning concentrate that is suitable for many common sample types
  • For some needle obstructions, mechanical cleaning may be necessary
For this purpose various sized tungsten cleaning wires can be used. They can be threaded through the needle to clear a blockage.
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally the solvent would be non-alkaline, non-phosphate, and non-detergent based. This is a non-phosphate, cleaning concentrate that is suitable for many common sample types.
Lieferumfang: The kit contains six different packs of wires and 70 ml cleaning concentrate.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge