TLC education advanced kit, micro-set F2
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
814300 814301 814302
552-0288EA 177 EUR InStock
552-0288 MANA814301 MANA814302
TLC education advanced kit, micro-set F2
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Edible fats
  • Fats and cholesterol in blood
Together with the materials from TLC micro-Set M samples of butter, margarine, and edible oil as well as blood can be separated and visualised by molybdatophosphoric acid spray reagent.
Achtung! Bitte beachten: * These products contain harmful substances which must be specially labelled as hazardous.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge