TSKgel® gel filtration columns, SW-type
Lieferant: TOSOH Bioscience
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0005102 0005103 0005104 0005146 0005147 0005148 0005371 0005758 0005788 0005789 0005790 0006727 0006728 0006729 0008540 0008541 0008542 0008801 0008805 0016049 0016215 0018762 0020025 0020027 0021485 0021845 0018674 0022855 0022854 0022856 0023448 0023450 0023449 0023451 0018675 0022857 0022859 0020026 0008543 0008800 0018008
554-0891EA 3540 EUR InStock
554-0891 554-0892 554-0893 554-0894 554-0895 554-0896 554-1605 554-1606 554-0897 554-0898 554-0899 554-0900 554-0901 554-0902 554-0903 554-0904 554-0905 554-0908 554-5604 554-5605 554-5606 554-1649 554-5609 554-5610 554-5611 554-5612 554-5607 554-5614 554-5613 554-5615 554-5317 554-5319 554-5318 554-5320 554-5608 554-5616 554-5617 TOYO0020026 554-1620 TOYO0008800 TOYO0018008
TSKgel® gel filtration columns, SW-type
Chromatographiesäulen
TSKgel® SW size exclusion columns are widely used for aqueous analysis and isolation of proteins, antibodies, enzymes, nucleic acids and other biological macromolecules.
  • Columns show high resolution and sensitivity and have excellent solvent stability with up to 100% polar organic solvents
  • The smaller particle size of the UltraSW and SWXL materials result in reduced analysis times
  • UP-SW columns are now available for UHPLC-SEC applications improving productivity and resolution
  • All SW-type columns are available in stainless steel hardware; SWXL columns are also available in PEEK hardware
  • TSKgel® SuperSW mAb HR is the ideal scouting column for unknown molecular weight compounds
TSKgel® UP-SW:
With modern advances in technology, Tosoh Bioscience have launched a range of 2 µm UHPLC columns for use in SEC. The TSKgel® UP-SW3000 columns are available in 150 and 300 mm lengths for use on modern, low dead volume HPLC or UHPLC instrumentation. They provide short analysis times and allow analysts to achieve improved resolution compared with the equivalent larger particle size columns. Offering the same exclusion range as the TSKgel® G3000SWXL columns, these columns are ideal for fast separations of monoclonal antibody analysis or can be used to improve your separation.
TSKgel® UltraSW:
Launched in 2013, TSKgel® UltraSW columns contain spherical 3 µm particles (300 Å pore size). TSKgel® UltraSW Aggregate has a slightly larger pore size than the TSKgel® SuperSW mAb columns and provides a wider separation window for higher order monoclonal antibody aggregates.
TSKgel® SuperSW:
In 1997, Tosoh Bioscience launched the initial TSKgel® SuperSW columns which contain spherical 4 µm particles in 4,6×300 mm dimensions and extended this portfolio in 2013 with further columns including the SuperSW mAb HTP suitable for UHPLC applications. This range offers high sensitivity even with limited sample mass. The TSKgel® SuperSW mAb HR - 'HR' indicating high resolution - delivers superior resolution over the whole molecular weight range, from fragments to aggregates, when analysing monoclonals. It has the same column dimensions and molecular weight range as the well-established TSKgel® G3000SWXL.
TSKgel® SWXL:
Introduced in 1987, the TSKgel® SWXL column line were the second generation TSKgel® phases developed with 5 or 8 µm particle size packings. The smaller particle sizes of the TSKgel® SWXL columns mean these can be implemented to improve sample resolution or to reduce analysis time (over SW columns). The G3000SWXL has proven to be the industry standard for QC analysis of monoclonal antibodies. Columns feature high pore volume per unit column volume, low sample adsorption and high column efficiency, all contributing to excellent sample resolution.
TSKgel® SW:
The TSKgel® SW column line, introduced in 1977, were the first of a long line of high-performance Gel Filtration (GFC) columns that have become synonymous with analysing protein molecular weights. It features 10 or 13 µm particle sizes with improved sample capacity for protein isolation due to the large pore sizes.
Zubehörinformationen: Corresponding guard columns are avaliable to protect the analytical columns. Direct connection, no holders are required.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge