VHP-Fittings für sehr hohen Druck, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHP-200 VHP-205 VHP-320X VHP-325 VHP-320 VHP-305 VHP-325X VHP-3200
554-1821EA 48.7 EUR InStock
554-1821 554-1823 554-5696 554-1997 554-1996 554-1995 554-5697 554-2270
VHP-Fittings für sehr hohen Druck, Upchurch Scientific®
Fittings und Verbinder
Reusable fittings made from stainless steel and proprietary PEEK polymer blend. Reusable up to 10 cycles without tubing damage or impact on pressure holding ability or carry-over (5 cycles if tightened to 1,6 N.m). Pressure rating up to 30 000 psi (2070 bar). Required torque 10 in-lbs (1,10 N-m)/14 in-lbs (1,6 N-m).
Stainless steel fittings have a double compression ferrule design. Pressure rating up to 30 000 psi (2070 bar). Required torque 20 in-lbs (2,25 N-m).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge