Protein based chiral HPLC columns, CHIRALPAK® AGP
Lieferant: CHIRAL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30713 30783 30781 30792 30793 30791 30784 30782 30734 30733 30714 30712 30794 30711
554-2472EA 1300 EUR InStock
554-2472 554-2476 554-2474 554-2479 554-2480 554-2478 554-2477 554-2475 554-4062 554-4061 554-2473 554-2471 554-2481 554-2470
Protein based chiral HPLC columns, CHIRALPAK® AGP
Chromatographiesäulen
These protein-based columns are suitable for separation of basic, acidic and non-protolytic compounds. These columns function entirely in the reversed phase chromatographic mode, using buffers with low organic modifier content and at moderate pH values.
  • Extremely broad applicability for separation of chiral drugs
  • Possibility to dynamically improve enantioseparation
  • Separates enantiomers of an extremely broad range of drug compounds such as amines, acids and non protolytes
CHIRALPAK® AGP is the second generation chiral separation column based on the use of α1-acid glycoprotein (AGP) as the CSP. Through a patented process, α1-AGP has been immobilised on porous, spherical silica particles (5 μm). The surface chemistry of the silica proves a stable chiral separation material with extremely broad applicability.
Racemic amines, acids and non protolytic compounds can be resolved directly, without derivatisation. The column enables resolution of a very large number of chiral compounds from different compound classes. This is due to the unique nature of the CSP, and the fact that enantioselectivity can be induced by choosing a proper mobile phase composition.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge