Reversed phase chromatography columns, RSpak DM-614
Lieferant: Resonac
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F7001002 F6700160
554-2916EA 1034 EUR InStock
554-2916 554-2873
Reversed phase chromatography columns, RSpak DM-614
Chromatographiesäulen
These columns are suitable for the analysis of amino acids and water-soluble vitamins.
  • Base material is polyhydroxymethacrylate
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge