HPLC columns, Cogent C8 300™
Lieferant: MICROSOLV
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40208-05P-2 40208-10P-1 40208-10P-2 40208-15P-1 40208-15P-2 40008-05P 40008-05P-1 40008-05P-2 40008-05P-2-3M 40008-10P 40008-10P-1 40008-10P-2 40008-10P-2-3M 40008-10P-3M 40008-15P 40008-15P-1 40008-15P-2 40008-15P-2-3M 40008-15P-3M 40008-25P 40008-25P-3M 40008-HG5 40008-HG6 40008-P21-100 40008-P21-150 40008-P21-250 40008-P21-50 40008-P30-100 40008-P30-150 40008-P30-250 40008-P30-50 40008-SP100 40008-SP150 40008-SP250 40008-SP250-3M 40008-SP50
554-6437EA 1120 EUR InStock
554-6437 554-6440 554-6441 554-6444 554-6445 554-6102 554-6103 554-6104 554-6105 554-6108 554-6109 554-6110 554-6111 554-6112 554-6115 554-6116 554-6117 554-6118 554-6119 554-6122 554-6126 554-6140 554-6142 554-6149 554-6150 554-6151 554-6152 554-6154 554-6155 554-6156 554-6157 554-6158 554-6159 554-6160 554-6161 554-6162
HPLC columns, Cogent C8 300™
Chromatographiesäulen
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
  • Extremely long column life
  • Polar and non-polar analytes
  • Increase precision of your results
  • Based on silica hydride
  • Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge