HPLC columns, Cogent Bidentate C18™
Lieferant: MICROSOLV
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40218-05P-2 40218-10P-2 40218-15P-1 40218-15P-2 40018-05P 40018-05P-1 40018-05P-2 40018-10P 40018-10P-1 40018-10P-2 40018-15P 40018-15P-1 40018-15P-2 40018-25P 40018-HG5 40018-HG6 40018-P21-100 40018-P21-150 40018-P21-250 40018-P21-50 40018-P30-150 40018-P30-250 40018-P30-50 40018-SP100 40018-SP150 40018-SP250 40018-SP50
554-6461EA 1130 EUR InStock
554-6461 554-6465 554-6468 554-6469 554-6176 554-6177 554-6178 554-6181 554-6182 554-6183 554-6186 554-6187 554-6188 554-6191 554-6201 554-6202 554-6208 554-6209 554-6210 554-6211 554-6214 554-6215 554-6216 554-6218 554-6219 554-6220 554-6221
HPLC columns, Cogent Bidentate C18™
Chromatographiesäulen
Using proprietary bonding technology, the straight chain C18 hydrocarbon is bonded directly to the TYPE-C silica-hydride surface at two separate points of attachment with direct siliconcarbon bonds. This Si-C bond is virtually indestructible and confers great stability and longevity to these columns.
  • Suitable for use with mobile phases containing from 100% water to 100% organic solvent
  • Very hydrophobic phase
  • Columns operate in RP, ONP, and ANP
  • Easy gradient analysis
These columns are ideal for generic or USP methods which specify the use of a C18 (or USP L1) column.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge