Capillary column end caps
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
260EC111
555-9468EA 80.4 EUR InStock
555-9468
Capillary column end caps
Fittings und Verbinder
These capillary column end caps are universal and fit any capillary column.
  • Reusable
  • Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
  • Colour coded to ensure that the column is reinstalled the same way it came out
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge