Nitrogen generator, Infinity 1051
Lieferant: PEAK SCIENTIFIC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
13-1282
564-0283EA 0 EUR InStock
564-0283
Nitrogen generator, Infinity 1051
Gasgeneratoren Stickstoff-Generatoren
Designed and engineered as an on-site, compressor less nitrogen generation solution tailored for the Shimadzu LCMS-8045, 8050 or 8060. The Infinity 1051 delivers one output of nitrogen and another of dry air to laboratories that already have an existing air supply.
  • Both a nitrogen and dry air output suitable for the Shimadzu LCMS-8045/50/60 and instruments requiring similar specifications
  • 24/7 operation at optimum performance if required
  • Gas is supplied on demand so generator works to your schedule
  • Few moving parts means little maintenance required and ensures long life of the generator
  • Minimal set-up required
  • Completely silent in operation
  • 1 year comprehensive on-site warranty
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge