Nitrogen gas generator, Genius SQ 24
Lieferant: PEAK SCIENTIFIC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3301661
564-0285EA 14770 EUR InStock
564-0285
Nitrogen gas generator, Genius SQ 24
Gasgeneratoren Stickstoff-Generatoren
For laboratories using any single quad LC-MS systems, the Genius SQ 24 nitrogen gas generator delivers analytical grade nitrogen supply dedicated for this instrument. With a compact size to fit under most laboratory benches and flow rates of up to 24 L/min, meeting and exceeding the maximum flow rate required by any Single Quad LC-MS on the market today, your laboratory can enjoy hassle free nitrogen gas which helps deliver the best analytical results.
  • Best value nitrogen generator on the market
  • Variable flow up to 24 L/min
  • Outlet pressure up to 8 bar (at 22 L/min maximum)
  • Reduced height and compact size to fit under most laboratory benches
  • Integrated internal compressors for easy plug and play
  • Based on proven and highly reliable technology
  • Backed by [Peak Protected] global on-site servicing, technical support and partner training
Genius SQ 24 has been designed using Peak’s extensive experience of working with and developing the best nitrogen generator membranes using expert suppliers and highly skilled Peak design and manufacturing engineers. This rigorous design process has helped not only deliver a reliable, high performance gas generator, it is also provides excellent value.
Zertifizierungen: CE marked
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge