di-Natriumhydrogenphosphat ≥99,7%, nach Ph. Eur. getestet, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
56814-1KG
56814-1KGEA 100.27 EUR InStock
56814-1KG
di-Natriumhydrogenphosphat ≥99,7%, nach Ph. Eur. getestet, Fluka™
di-Natriumhydrogenphosphat
Formel: Na₂HPO₄
Melting Pt: 250 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003496
CAS-Nummer: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805
Order Now
Spezifikation Testergebnisse
pKa (25 °C) (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
Identity A complying
Identity C complying
Identity D complying
Assay (calculated to the dried substance) 98.0 - 101.0 %
Assay of NaH₂PO₄ (calculated to dried substances) Max. 2.5 %
Appearance of the solution complying
Loss on drying (105°C, 4 h) Max. 1.0 %
Arsenic (As) Max. 2 ppm
Iron (Fe) Max. 20 ppm
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 200 ppm
Sulfate (SO₄) Max. 500 ppm
Reducing impurities complying
Residual solvents complying
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge