Supelco® Discovery® Cyano HPLC Columns
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
59356-U30 59357-U40
59356-U30EA 786 EUR InStock
59356-U30 59357-U40
Supelco® Discovery® Cyano HPLC Columns
Chromatographiesäulen
Discovery® Cyano columns have low hydrophobicity for rapid elution of hydrophobic analytes with exceptional peak shape for basic and acidic analytes.
  • Suitable for LC/MS application
  • High retention and resolution of polar compounds
  • Compatible with 100% aqueous phases
Stable pH: 2,0 to 8,0
Lieferumfang: Validation packs includes three columns, each from a different lot of bonded phase. Supelguard™ guard cartridge kit includes one cartridge, a stand-alone holder, a piece of tubing, two nuts and ferrules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge