Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60137-250ML 60137-1L
60137-250MLEA 27.6 EUR InStock
60137-250ML 60137-1L
Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Elektrolyte Elektrodenfülllösungen
~3 M Potassium chloride solution saturated with silver chloride (~0,16 M AgCl).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge