Analogue stopwatch, Mega Minute
Lieferant: HANHART
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
185.7901-00
609-0148EA 592 EUR InStock
609-0148
Analogue stopwatch, Mega Minute
Timer Stoppuhren, analog
Stopwatch with sprung central minutes hand for measuring up to 60 minutes.
  • Chrome plated and polished brass casing, diamond cut
  • 1/5 second recorder
  • Excellent readability thanks to double-sized minute scale in contrasting colour, and sprung minute hand
  • Precision lever movements (7 jewels)
  • Shock protection, additional dust and water protection
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge