VWR® Traceable® Four-Channel Timer
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
609-0234EA 41.4 EUR InStock
609-0234
VWR® Traceable® Four-Channel Timer
Timer Timer, digital
Calibrated timer and digital clock is always near-at-hand with its clip-on, magnet, and benchtop configurations with adjustable viewing angle.
  • Ergonomic design provides optimal viewing angle, easy use, and increased stability on bench
  • Antimicrobial protection is built in for a cleaner, more hygienic product
  • Large, tactile buttons are easy to use when wearing gloves
  • This timer steps in time with 4 channels to time multiple events at once and default digital clock function
  • Maintain compliance; includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Repetitive or recurring tasks are quick and easy with on-board memory function that returns display to previously programmed countdown time
  • Never miss your cue with extra-loud alarms that sound for 1 full minute
  • LCD display
This timer allows you to count up to or down from 99 hours, 59 minutes, 59 seconds with 1-second resolution. This timer features a ³/₄" high display with four channels that can be used simultaneously. Each channel has a distinctive alarm that sounds for 1 minute. Pressing any button will turn off the alarm. Timer memory recalls previous time set. Features time of day in 12- or 24-hour formats.
Zertifizierungen: NIST Traceable.
Lieferumfang: Includes timer, AAA battery, quick start guide, and Traceable® certificate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge