Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Di-8-ANEPPS Membranfarbstoff, für die Histologie

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


61012
61012.EA 536 EUR InStock
61012.
Di-8-ANEPPS Membranfarbstoff, für die Histologie
Di-8-ANEPPS
ANEPPS dyes are fast responding membrane potential dyes. Compared to Di-4-ANEPPS, Di-8-ANEPPS is better retained in the outer leaflet of cell plasma membranes and thus more suitable for long-term membrane potential studies. Di-8-ANEPPS is also more photostable and less phototoxic than Di-4-ANEPPS

  • λEx/λEm (MeOH) = 498/713 nm*
  • Orange red solid soluble in DMF, ethanol or DMSO
  • Store at 4°C and Protect from light, especially when in solution

Biotium offers both Di-4-ANEPPS and Di-8-ANEPPS. Di-4-ANEPPS has the problem of being rapidly internalized in cells, so it is mainly used for short-term experiments. However, because Di-8-ANEPPS is more lipophilic, the dye is better retained in the outer leaflet of cell plasma membranes and thus more suitable for long-term membrane potential studies. Di-8-ANEPPS is also more photostable and less phototoxic than Di-4-ANEPPS.

* Note that the spectra of styryl dyes generally undergo a large blue shift in lipids compared to organic solvent. See the spectra for styryl dyes in liposomes under product images.


Formel: C₃₆H₅₂N₂O₃S
Storage Temperature: Kühlschrank
MDL Number: MFCD03453608
CAS-Nummer: 157134-53-7
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge