RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt] Membranfarbstoff, für die Histologie
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
61018
61018.EA 326 EUR InStock
61018.
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt] Membranfarbstoff, für die Histologie
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
The styryl dye RH237 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. Excitation/emission data (below) is for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission.
  • λEx/λEm (MeOH) = 528/782 nm
  • Dark solid soluble in DMSO
  • Store at -20°C and protect from light
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6369d19cd73fda9552b83eee81e0b78167d38faf&vid=6369d19cd73fda9552b83eee81e0b78167d38faf&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge