Infrared moisture analyser, MA35
Lieferant: Sartorius Balances
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MA35M-230N
611-4357EA 2940 EUR InStock
611-4357
Infrared moisture analyser, MA35
Analysatoren Feuchte-Analysatoren
This infrared moisture analyser is designed for use in daily routine in-process control.
  • Uniform distribution of the heat rays using an integrated reflector
  • Heating of a sample by two metal tubular-shaped heating elements
  • Uniform distribution of the heat rays using an integrated reflector
Quick and reliable analysis of the material moisture of liquid, pasty and solid substances using the thermogravimetric method.
Zertifizierungen: FDA/HACCP regulations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge