Mikrowaage SECURA 26
Lieferant: Sartorius Balances
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SECURA26-1S
611-5067EA 23140 EUR InStock
611-5067
Mikrowaage SECURA 26
Waagen
The Secura® microbalance provides highly accurate weighing results and operating convenience, and also features built-in protection systems for complete reliability and regulatory compliance. This microbalance has a readability of 2 microgrammes.
  • Integrated protection systems minimise the risk of obtaining incorrect results
  • Real-time alarm messaging and interactive user guidance prevent inaccurate results due to an unlevelled balance
  • SQmin for automatic detection and active monitoring ensure you are within the acceptable USP operating range for reliable results
  • Fully automatic internal adjustment function, isoCAL, for the highest possible accuracy at all times
  • Cal Audit Trail for easy, traceable documentation of all calibration and adjustment data
  • Safe and clear documentation with sample and batch identifiers for keeping track of weighing data
  • Standard password security features give protection against tampering and unauthorised access
Echtzeit-Bedienerführung beim Nivellieren der Waage, interne vollautomatische Justierautomatik sowie klar rückführbare Dokumentation mit Proben- und Chargen-Identnummern machen Ihre Arbeit im Labor effizienter. Einfache Aufzeichnungen mit der integrierten PC-Direkt-Funktion und der Plug & Work-Technologie für den Anschluss an einen Sartorius Labordrucker.
Lieferumfang: Supplied with universal power adapter, in-use cover, application guide, draught ring.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge