Dishes, disposable
Lieferant: Korff
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60505 60530 60540 60545 60550 60560 60558 60500 60510 60520 60525 60552
KORF60505EA 48.7 EUR InStock
KORF60505 KORF60530 KORF60540 KORF60545 KORF60550 KORF60560 KORF60558 611-9031 611-9032 611-9033 611-9034 611-9035
Dishes, disposable
Schiffchen Wägeschiffchen
Aluminium.
  • Lightweight
  • Good chemical resistance
  • Good temperature resistance
Weighing/drying pan, oil-free, ideal for general purpose weighing, dispensing, storage and drying.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge