Weighing paper, grade MN 226
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186002
611-9091EA 14 EUR InStock
611-9091
Weighing paper, grade MN 226
Wägepapier
This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that the weighed goods can be transferred without loss.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge