Burettes, straight glass stopcock, class A, Volac®
Lieferant: Poulten & Graf
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RB9803/WAC/I
612-1694EA 81.26 EUR InStock
612-1694
Burettes, straight glass stopcock, class A, Volac®
Büretten
Burette, Class A, made of borosilicate glass, stopcock with interchangeable glass key.
  • Clearly legible lettering and fine graduations lines in white color
  • Wide hopper for easy filling
  • Interchangeable glass spikes for better association
Class A burettes are individually calibrated, tested at 3 points
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge