Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Lieferant: GILSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F161401
612-1816EA 132 EUR InStock
612-1816
Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Pipettenständer
This carousel stand holds up to seven single or multi channel Gilson pipettes.
  • Robust and stable due to its heavy base
  • Made of PP and PC: Strong resistance to aggressive chemicals
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives users an easy access to the pipette of their choice
  • Space saving for a a neat, tidy and organised work bench
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge