Messkolben, Klasse A, weiße Graduierung, Werkszertifikat
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3151/04WC 3151/06WC 3151/08WC 3151/10WC 3151/12WC 3151/14WC 3151/16WC 3151/18WC 3151/20WC 3151/22WC 3151/24WC 3151/26WC 3151/01WC 3151/02WC
612-3232EA 155 EUR InStock
612-3232 612-3239 612-3202 612-3237 612-3234 612-3220 612-3214 612-3199 612-3229 612-3227 612-3225 612-3223 612-3200 612-3201
Messkolben, Klasse A, weiße Graduierung, Werkszertifikat
Kolben Messkolben
PYREX®, Borosilikatglas, mit PE-Stopfen.
  • Calibrated to the tolerances required by the United States Pharmacopoeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with individual serial number and date of manufacture
  • Calibrated to contain (TC, In)
  • White enamel inscriptions
Complies with ISO 1042 and DIN 12664.
Bestellinformation: Supplied with certificate of calibration traceable to National Standards.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge