Messkolben, Klasse A, weiße Graduierung
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3152/08A 3152/10A 3152/14A 3152/16A 3152/20A 3152/04A 3152/06A 3152/12A 3152/18A 3152/22A
612-3248EA 92.2 EUR InStock
612-3248 612-3251 612-3244 612-3243 612-3247 612-2903 612-2904 612-3249 612-3242 612-3245
Messkolben, Klasse A, weiße Graduierung
Kolben Messkolben
PYREX®, Borosilikatglas, braun, mit PE-Stopfen.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • White enamel inscriptions, ideal for use with light sensitive solutions
Zertifizierungen: BS EN ISO 1042, DIN 12664
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge