Volumetric flasks, class B, blue markings
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FRD022P FRD024P FRD025P FRD032P FRD040P FRD026P FRD028P FRD030P FRD034P FRD036P FRD038P
612-3329EA 56.1 EUR InStock
612-3329 612-3330 612-3331 612-3335 612-3336 612-3332 612-3333 612-3334 BIBBFRD034P BIBBFRD036P BIBBFRD038P
Volumetric flasks, class B, blue markings
Kolben Messkolben
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Excellent temperature resistance
Zertifizierungen: Complies with ISO 1042 and DIN 12664
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge