Bulb pipettes, class B, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
243370108 243370202 243370708 243370802 243371207 243371404 243371704 243372409
612-5234EA 7.9 EUR InStock
612-5234 612-5235 612-5236 612-5237 612-5238 612-5239 612-5240 612-5241
Bulb pipettes, class B, DURAN®
Pipetten Vollpipetten
Soda-lime glass (AR-Glas®).
  • Marks and inscription in brown
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 648.
Colour code complies to DIN 12621.
*no bulb
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge