VWR® Pipette Filler, White/Blue
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
612-7284EA 54.7 EUR InStock
612-7284
VWR® Pipette Filler, White/Blue
Pipetten Pipettierhelfer
The VWR® pipette filler provides precise fluid control at your fingertips.
  • A T-bar thumb lever controls the aspirating and dispensing speeds from serological pipettes from 1 to 100 ml in size
  • Conveniently accommodates either glass or plastic serological or volumetric pipettes ranging from 1 to 100 ml
  • Includes a replaceable 0,45 μm hydrophobic filter to protect your samples and internal components
  • Inline ergonomic design reduces stress with ease of handling and operation
Comfortable to hold and easy to use, the manual inline pipette controller was designed with an ergonomic grip and bulb for simple and precise operation.
Body material: ABS
Bestellinformation: For replacement filters; please contact your Avantor representative.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge