Syringes, 3-piece, HENKE-JECT®
Lieferant: HENKE SASS WOLF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8300018745
613-2793EA 43.3 EUR InStock
613-2793
Syringes, 3-piece, HENKE-JECT®
Spritzen Spritzen, medizinisch
These syringes are commonly used for embryo transfer and oocyte handling.
  • Rubber piston syringes
  • Double seal for increased safety
  • Pyrogen-free, PVC-free, non-toxic
Syringes have smooth flow, tight and high transparency barrel and the lubricated synthetic rubber piston allows fine dosing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge