BD Discardit™ II Syringes
Lieferant: BECTON DICKINSON MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
300296 300928 309040 309050 309110 300322 300512 300199 300296UK 300512UK 301360
613-3916EA 30.2 EUR InStock
613-3916 613-3932 BDAM309040 613-3951 613-3952 615-0031 615-0032 BDAM300199 BDAM300296UK BDAM300512UK BDAM301360
BD Discardit™ II Syringes
Spritzen Spritzen, medizinisch
The BD Discardit™ II two-piece syringe is a single-use medical device intended for injection and aspiration of medical fluids, including both bodily fluids (blood, etc.) and medicines. The lubricant included in the barrel material allows smooth advancement of the plunger, leading to reduced sliding force.
  • Clear and transparent barrel for good visualisation of syringe content
  • Leak-tight
  • Accurate graduation suited to user's needs
  • Robust retaining ring to prevent accidental plunger rod pullout during aspiration
  • Design of finger grip flanges ensures stability and comfort during injection
  • Ergonomically designed plunger for single-handed operation
  • Sterile, single-use
  • Natural rubber latex is not part of the material formulation
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge