Charging pipette carousel for four pipettes
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
730991
613-4124EA 646 EUR InStock
613-4124
Charging pipette carousel for four pipettes
Pipettenständer
The charging carousel for four pipettes is a convenient, easy-to-use storage and charge system designed to keep your electronic pipettes ready for use and safe from contamination.
  • Charging carousel has a simple, elegant design
  • Holds pipettes in an upright position while not in use to avoid contamination from benches and tables
  • Compact carousel stand: Saves valuable bench space
Proper use and storage will ensure long-lasting use of your electronic pipettes, keeping them ready for whenever you need them. When storing your electronic pipette (or any pipette), it should be stored in an upright position within the stand. Storing in this manner helps ensure cleanliness and safety while avoiding contamination from things like desks and benches. It is recommended that electronic pipettes be stored and charged on a charging carousel whenever they are not in use. In this way, the batteries always remain charged, ready for use whenever work begins. The Charging carousel was designed with compact storage needs in mind.
As more vital technology and products become available for improving workflows, streamlining applications and keeping up with demand, it is vital to ensure your workspace is efficiently organised. These compact carousels are ideal for saving bench space and ensuring a safe and clean environment. Regardless of the pipette (e.g., channel number) being used, the Charging Carousel can help usage of your Picus® electronic pipettes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge