Pipette stand, linear
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
725620
613-8706EA 84 EUR InStock
613-8706
Pipette stand, linear
Pipettenständer
This pipette stand holds pipettes in an upright position while not in use to avoid contamination from benches and tables. Designed for all Sartorius mechanical and electronic pipettes, particularly for mLINE®, Proline® Plus and Proline® mechanical devices. Also compatible with other brands.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge