Universal water bath, analogue, WB 436-A
Lieferant: GERBER FUNKE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3708
614-2063EA 677.5 EUR InStock
614-2063
Universal water bath, analogue, WB 436-A
Bäder Wasserbäder, ohne Umwälzung
Analogue water bath with stainless steel inner and outer casing. Heating elements are located separately within the casing. Ideal for dairy use, wide range of accessories available. It is recommended to use distilled water with the bath.
  • Analogue temperature control with adjusting knob
  • PT100 sensor
  • Thermostatic heat controller
  • Overheating protection (even when bath is empty)
Lieferumfang: Includes thermometer with temperature display.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge