Automatic viscometers, ViscoSystem® AVS® 470
Lieferant: SI Analytics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
285415709 285415708
616-0026EA 12370 EUR InStock
616-0026 616-1115
Automatic viscometers, ViscoSystem® AVS® 470
Viskosimeter Automatische Kapillarviskosimetersysteme
The ViscoSystem AVS 470 is the first measuring device that allows suction and pressure measurements completely independent of a PC.
  • Determination with pressure or with vacuum for high viscosities
  • Both detections methods optical or thermal are possible
  • Nearly all necessary calculations are integrated
  • Additional parameters can be set up over a regular PS 2 keyboard
This brings maximum independence and flexibility, allowing you to set up a measuring station, that meets highest requirements, even under difficult conditions, to monitor production or control quality in the polymer and mineral oil industry.
The ViscoSystem AVS 470 determines the time which the sample to be examined requires to flow through the measuring distance in the capillary viscometer with quartz precision.
All common types of viscosity calculations are already integrated into the device, a small PS 2 keyboard is all you need to enter additional data.
A serial printer can be used to conveniently document your measuring results.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge