Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
61768-1ML
61768-1MLEA 225 EUR InStock
61768-1ML
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
Protein-Aufbereitungs Protein Purification Resins
50% aqueous suspension in 1 M NaCl.
  • Capacity: 0,1 to 1,0 mg/ml, packed gel capacity (ovomucoid)
Application: Protein isolation from bordetella pertussis that facilitates entry of calmodulin-sensitive adenylate cyclase into animal cells.
Achtung! Bitte beachten: Storage temperature: Refrigerator.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge