Active gauge displays
Lieferant: EDWARDS HIGH VACUUM METTLER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D39565000 D39566000 D39567000 D39585000 D39561000
619-0223EA 0 EUR InStock
619-0223 619-0224 619-0225 EDWAD39585000 EDWAD39561000
Active gauge displays
Pumpen Vakuum-Messgeräte
The active gauge displays are a range of simple, low cost units for the measurement and control of vacuum from atmosphere to 10-9 mbar.
  • Single switch with LED indication for gauge head selection
  • Option of two adjustable set-points with relay status indication
  • Mains power input supply via IEC connector
  • Clear analog display covers the whole gauge head pressure range
  • Available with both bench top use or panel mounting
Lieferumfang: Supplied with tilt feet for convenient bench top mounting and panel mounting kit to DIN 43700 standard.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge