Temperature chart recorders, disposable
Lieferant: DICKSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D302
620-0432EA 149 EUR InStock
620-0432
Temperature chart recorders, disposable
Diagrammschreiber Diagrammschreiber
These are single use logger accurately records temperatures from start to finish of transport and documents adherence to critical storage environments.
  • Failsafe start-up and pull out start tab
  • Tamper-proof crimped seal
  • Shock and vibration protection
Lieferumfang: Supplied with a shipment/job sheet and a mounting tape.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge