Thermometer clamp, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F182410000
620-1030EA 48.8 EUR InStock
620-1030
Thermometer clamp, SP Bel-Art
Klemmen Klemmen für den allgemeinen Gebrauch
These PP clips hold thermometers and probes from 3,2 to 11 mm in diameter securely in place.
  • Fits onto the rims of beakers or pitchers up to ¼” thick
  • Autoclavable at 121 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge