Chart for vaccine chart recorder, 4"
Lieferant: DICKSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C204
620-1484EA 114 EUR InStock
620-1484
Chart for vaccine chart recorder, 4"
Diagrammschreiber Diagrammscheiben für Schreiber
The vaccine chart recorder can be placed right inside the refrigerator or freezer along with the vaccines.
  • Easy-to-read special vaccine chart with circular bands highlight the acceptable vaccine ranges of 2 to 8 °C
  • LCD display shows current temperature for quick reference
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge