VWR® RM 10, Moisture Tester
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
620-1956EA 180 EUR InStock
620-1956
VWR® RM 10, Moisture Tester
Analysatoren Feuchte-Analysatoren
The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or wash floor down to 4 cm.
  • Easy and fast moisture rating, non destructive measurement
  • Displayed values by „digit“ are relative so that values can be compared
  • To check moisture on surfaces – No injection into the measuring object required
Suitable for high-speed determination of moisture i.e. estate agents, property management, house owners, architects, building experts, building contractors.
Achtung! Bitte beachten: This is an indicator for fast estimation – Does not replace precision instruments like the RM 20.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge